Assay Details
Target Gene Details
Entrez Gene ID: | 21752 |
Gene Name: | telomerase reverse transcriptase |
Gene Aliases: |
EST2, TCS1, TP2, TR, TRT |
Location: |
Chr.13:73626797-73649843 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tert | NM_009354.1 | NP_033380.1 | ||
| AF051911.1 | AAC09323.1 | |||
| BC127068.1 |
More Information
Set Membership: |
|

Genomic Map