Assay Details
Target Gene Details
Entrez Gene ID: | 14423 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 1 |
Gene Aliases: |
- |
Location: |
Chr.18:24205344-24286816 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galnt1 | NM_001160404.1 | 3 | 336 | NP_001153876.1 |
| NM_013814.3 | 3 | 424 | NP_038842.3 | |
| AK083391.1 | 3 | 337 | ||
| BC056215.1 | 3 | 341 | AAH56215.1 | |
| BC090962.1 | 2 | 214 | AAH90962.1 | |
| BY711053.1 | 3 | 386 | ||
| U73820.1 | 1 | 89 | AAB58477.1 |
More Information
Set Membership: |
|

Genomic Map