Assay Details
Target Gene Details
Entrez Gene ID: | 353211 |
Gene Name: | prune homolog 2 (Drosophila) |
Gene Aliases: |
6330414G02Rik, A230083H22Rik, A330102H22Rik, Bmcc1, mKIAA0367 |
Location: |
Chr.19:16955670-17223958 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prune2 | NM_181348.4 | 12 | 8707 | NP_851993.3 |
AK038997.1 | 4 | 471 | BAC30198.1 | |
AK054190.1 | 4 | 470 | BAC35688.1 | |
AK163576.1 | 4 | 460 | BAE37405.1 | |
AK165582.1 | 4 | 523 | BAE38273.1 | |
AK170001.1 | 4 | 401 | BAE41506.1 | |
AK172939.1 | 4 | 460 | BAD32217.1 | |
BC094224.1 | 5 | 2951 | AAH94224.1 | |
BC095978.1 | 4 | 445 | AAH95978.1 | |
FR693373.1 | 4 | 469 | ||
GQ403051.1 | 4 | 462 |
