Assay Details
Target Gene Details
Entrez Gene ID: | 14913 |
Gene Name: | guanylate cyclase activator 1a (retina) |
Gene Aliases: |
GC-A, Gcap1, Guca1, mGCAP1 |
Location: |
Chr.17:47394558-47413514 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Guca1a | NM_008189.2 | NP_032215.2 | ||
| BC026834.1 | AAH26834.1 | |||
| BC031810.1 | AAH31810.1 | |||
| L36860.1 | AAA60716.1 |

Genomic Map