Assay Details
Target Gene Details
Entrez Gene ID: | 72167 |
Gene Name: | THUMP domain containing 2 |
Gene Aliases: |
2810025A12Rik |
Location: |
Chr.17:81026327-81065127 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Thumpd2 | NM_028138.1 | NP_082414.1 | ||
| AK012806.1 | BAB28484.1 |
More Information
Set Membership: |
|

Genomic Map