Assay Details
Target Gene Details
Entrez Gene ID: | 224530 |
Gene Name: | acetyl-Coenzyme A acetyltransferase 3 |
Gene Aliases: |
ACTL |
Location: |
Chr.17:12923959-12940396 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acat3 | NM_153151.3 | NP_694791.2 | ||
| AF356876.1 | AAM00222.1 | |||
| AK143884.1 | BAE25582.1 | |||
| AK149441.1 | BAE28878.1 | |||
| BC049873.1 | AAH49873.1 |
More Information
Set Membership: |
|

Genomic Map