Assay Details
Target Gene Details
Entrez Gene ID: | 83965 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 5 |
Gene Aliases: |
AW061004, D17Abb1e, E-NPP 5, mKIAA4152 |
Location: |
Chr.17:44078796-44086567 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Enpp5 | NM_001168620.1 | NP_001162091.1 | ||
NM_032003.2 | 1 | 51 | NP_114392.1 | |
AF233377.1 | AAG49143.1 | |||
AK032341.1 | ||||
AK220510.1 | BAD90517.1 | |||
BC089320.1 | AAH89320.1 | |||
BC138977.1 | ||||
BC138978.1 | ||||
BY232106.1 | ||||
BY252780.1 | 1 | 53 |
