Assay Details
Target Gene Details
Entrez Gene ID: | 70345 |
Gene Name: | RIKEN cDNA 0610038B21 gene |
Gene Aliases: |
- |
Location: |
Chr.8:77517056-77518578 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 0610038B21Rik | NR_028125.1 | 1 | 1044 | |
| AK002788.1 | 1 | 1045 | BAB22359.1 |

Genomic Map