Assay Details
Target Gene Details
Entrez Gene ID: | 69085 |
Gene Name: | zinc finger, CCHC domain containing 9 |
Gene Aliases: |
1810019C21Rik |
Location: |
Chr.13:91796520-91807997 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zcchc9 | NM_145453.2 | 6 | 1048 | NP_663428.2 |
AK044802.1 | 5 | 841 | BAC32099.1 | |
AK051610.1 | 4 | 2938 | ||
BB291225.2 | 1 | 179 | ||
BC024496.1 | 5 | 972 | AAH24496.1 |
