Assay Details
Target Gene Details
Entrez Gene ID: | 13682 |
Gene Name: | eukaryotic translation initiation factor 4A2 |
Gene Aliases: |
4833432N07Rik, BM-010, Ddx2b, Eif4, eIF-4A-II, eIF4A-II |
Location: |
Chr.16:23107468-23114132 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif4a2 | NM_001123037.2 | 10 | 2149 | NP_001116509.1 |
| NM_001123038.2 | 11 | 3105 | NP_001116510.1 | |
| NM_013506.3 | 11 | 2289 | NP_038534.2 | |
| NR_110335.1 | 12 | 2396 | ||
| AK165898.1 | 10 | 1860 |
Target Gene Details
Entrez Gene ID: | 106344 |
Gene Name: | replication factor C (activator 1) 4 |
Gene Aliases: |
A1, AI894123, AU040575, RFC37 |
Location: |
Chr.16:23113948-23127730 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rfc4 | NM_145480.1 | 11 | 1112 | NP_663455.1 |
| AK146954.1 | 11 | 1100 | BAE27563.1 | |
| AK147030.1 | 10 | 1179 | BAE27622.1 | |
| BC003335.1 | 11 | 1112 | AAH03335.1 |
More Information
Set Membership: |
|

Genomic Map