Assay Details
Target Gene Details
Entrez Gene ID: | 78185 |
Gene Name: | protein phosphatase 1, regulatory subunit 3F, opposite strand |
Gene Aliases: |
4930524L23Rik |
Location: |
Chr.X:7573600-7581016 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r3fos | NR_029473.1 | 3 | 716 | |
AK019695.1 | 3 | 522 | BAB31838.1 | |
AK048169.1 | 3 | 715 | ||
AK080141.1 | 3 | 497 | BAC37832.1 | |
AK080337.1 |
