Assay Details
Target Gene Details
Entrez Gene ID: | 170472 |
Gene Name: | RecQ protein-like 5 |
Gene Aliases: |
RecQ5, Recq5b, Recql5b |
Location: |
Chr.11:115892595-115933531 on Build GRCm38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Recql5 | NM_130454.2 | NP_569721.1 | ||
| AB059999.1 | BAB79232.1 | |||
| AK049269.1 | BAC33647.1 | |||
| AK050965.1 | BAC34479.1 | |||
| AK083513.1 |
Target Gene Details
Entrez Gene ID: | 66528 |
Gene Name: | small integral membrane protein 5 |
Gene Aliases: |
2210020M01Rik |
Location: |
Chr.11:115899512-115906269 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smim5 | NM_183259.3 | 3 | 518 | NP_899082.1 |
| AK004370.1 | 3 | 451 | BAE43175.1 | |
| AK020450.1 | 3 | 480 | BAC25633.1 |
More Information
Set Membership: |
|

Genomic Map