Assay Details
Target Gene Details
Entrez Gene ID: | 52635 |
Gene Name: | extended synaptotagmin-like protein 2 |
Gene Aliases: |
2410017M09Rik, 4921504I16Rik, CHR2SYT, D12Ertd551e, Fam62b |
Location: |
Chr.12:116279680-116390935 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esyt2 |

Genomic Map