Assay Details
Target Gene Details
Entrez Gene ID: | 70988 |
Gene Name: | RIKEN cDNA 4931428L18 gene |
Gene Aliases: |
4930563O14Rik |
Location: |
Chr.1:31141078-31222656 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4931428L18Rik | NR_033445.1 | 3 | 507 | |
AK016483.1 | 4 | 463 | BAB30263.1 |
