Assay Details
Target Gene Details
Entrez Gene ID: | 394430 |
Gene Name: | UDP glycosyltransferase 1 family, polypeptide A10 |
Gene Aliases: |
A13, Ugt1a13 |
Location: |
Chr.1:88055411-88220002 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ugt1a10 | NM_201641.2 | NP_964003.2 | ||
| AY227201.1 | AAP48600.1 | |||
| BC089569.1 | AAH89569.1 |
Target Gene Details
Entrez Gene ID: | 394434 |
Gene Name: | UDP glucuronosyltransferase 1 family, polypeptide A9 |
Gene Aliases: |
UDPGT, UDPGT 1-9, UGT1-09, UGT1.9, UGTP4, Ugt1a12, mUGTBr/P |
Location: |
Chr.1:88070779-88220002 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ugt1a9 | NM_201644.2 | 1 | 534 | NP_964006.2 |
| AY227200.1 | 1 | 513 | AAP48599.1 | |
| BC138699.1 | 1 | 513 | ||
| BC146021.1 | 1 | 513 | ||
| L27122.1 | 1 | 459 | AAA40524.1 | |
| S57479.1 | 1 | 459 | AAB26034.1 |
More Information
Set Membership: |
|

Genomic Map