Assay Details
Target Gene Details
Entrez Gene ID: | 14617 |
Gene Name: | gap junction protein, delta 2 |
Gene Aliases: |
Gja9, connexin36, cx36 |
Location: |
Chr.2:114009601-114013619 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gjd2 | NM_010290.2 | NP_034420.2 | ||
| BC058595.1 | AAH58595.1 |
More Information
Set Membership: |
|

Genomic Map