Assay Details
Target Gene Details
Entrez Gene ID: | 70163 |
Gene Name: | LY6/PLAUR domain containing 8 |
Gene Aliases: |
2210415F13Rik |
Location: |
Chr.11:58379032-58390544 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lypd8 | NM_001083884.1 | NP_001077353.1 | ||
| NM_027339.2 | NP_081615.2 | |||
| AK008940.1 | BAB25981.1 | |||
| BC026828.1 | AAH26828.1 |
More Information
Set Membership: |
|

Genomic Map