Assay Details
Target Gene Details
Entrez Gene ID: | 232016 |
Gene Name: | coiled-coil domain containing 129 |
Gene Aliases: |
D530004J12Rik, Gm458 |
Location: |
Chr.6:55804628-55978743 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc129 | NM_001081665.1 | NP_001075134.1 | ||
| AK085190.1 | BAC39386.1 | |||
| BC116335.2 |
More Information
Set Membership: |
|

Genomic Map