Assay Details
Target Gene Details
Entrez Gene ID: | 170770 |
Gene Name: | BCL2 binding component 3 |
Gene Aliases: |
PUMA, PUMA/JFY1 |
Location: |
Chr.7:16308321-16318334 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bbc3 | NM_133234.2 | NP_573497.1 | ||
| AF332560.1 | AAK31317.1 | |||
| BC044782.2 | AAH44782.1 | |||
| BC060370.1 | AAH60370.1 | |||
| BC147261.1 | ||||
| BC147262.1 |
More Information
Set Membership: |
|

Genomic Map