Assay Details
Target Gene Details
Entrez Gene ID: | 67384 |
Gene Name: | BCL2-associated athanogene 4 |
Gene Aliases: |
2410112I15Rik, SODD |
Location: |
Chr.8:25764538-25785209 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bag4 | NM_026121.3 | NP_080397.1 | ||
| AF332863.1 | AAL99586.1 | |||
| AK010765.1 | BAB27167.1 | |||
| AK136899.1 | BAE23161.1 | |||
| AK160316.1 | ||||
| AK162658.1 | BAE37009.1 | |||
| BC037239.1 | AAH37239.1 | |||
| BC058518.1 | AAH58518.1 | |||
| BC138201.1 | ||||
| BC145986.1 |
More Information
Set Membership: |
|

Genomic Map