Assay Details
Target Gene Details
Entrez Gene ID: | 14816 |
Gene Name: | glutamate receptor, metabotropic 1 |
Gene Aliases: |
4930455H15Rik, ENSMUSG00000075319, Gm10828, Gprc1a, mGluR1, nmf373, rcw, wobl |
Location: |
Chr.10:10686059-11082377 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grm1 | NM_001114333.2 | NP_001107805.1 | ||
| NM_016976.3 | NP_058672.1 | |||
| AF320126.2 | AAG41991.2 | |||
| BC067057.1 | AAH67057.1 | |||
| BC079566.1 | AAH79566.1 |
More Information
Set Membership: |
|

Genomic Map