Assay Details
Target Gene Details
Entrez Gene ID: | 218121 |
Gene Name: | membrane bound O-acyltransferase domain containing 1 |
Gene Aliases: |
9130215M02Rik, BC023845, LPEAT1, Moact1, Oact1 |
Location: |
Chr.13:30136449-30246694 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mboat1 | NM_153546.4 | NP_705774.1 | ||
| AB297382.1 | ||||
| AB305045.1 | ||||
| AK033388.1 | BAC28261.1 | |||
| AK137755.1 | ||||
| AK146984.1 | BAE27587.1 | |||
| BC023845.1 | AAH23845.1 | |||
| BC024653.1 | AAH24653.1 |
More Information
Set Membership: |
|

Genomic Map