Assay Details
Target Gene Details
Entrez Gene ID: | 66591 |
Gene Name: | MAD2L1 binding protein |
Gene Aliases: |
0610009D16Rik, 2510031P20Rik, AU018945, AW550977, C76045, CMT2A, CMT2B, Mad2lbp, mKIAA0110 |
Location: |
Chr.17:46147385-46153551 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mad2l1bp | NM_025649.3 | NP_079925.2 | ||
| AK002377.1 | BAB22054.1 | |||
| AK004224.1 | BAB23226.1 | |||
| AK007974.1 | ||||
| AK156569.1 | BAE33758.1 | |||
| AK220322.1 | BAD90394.1 | |||
| BC005768.1 | AAH05768.1 |
More Information
Set Membership: |
|

Genomic Map