Assay Details
Target Gene Details
Entrez Gene ID: | 67397 |
Gene Name: | endoplasmic reticulum protein 29 |
Gene Aliases: |
1200015M03Rik, 2810446M09Rik, AW209030, Erp28, Erp31, PDI-Db |
Location: |
Chr.5:121444753-121452474 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erp29 | NM_026129.2 | NP_080405.1 | ||
| AB009293.1 | ||||
| AK004795.1 | BAB23570.1 | |||
| AK009881.1 | BAB26559.1 | |||
| AK013303.1 | ||||
| AK154539.1 | BAE32664.1 | |||
| AK168112.1 | BAE40083.1 | |||
| BC017125.1 | AAH17125.1 |
More Information
Set Membership: |
|

Genomic Map