Assay Details
Target Gene Details
Entrez Gene ID: | 66935 |
Gene Name: | corepressor interacting with RBPJ, 1 |
Gene Aliases: |
1700023B02Rik, 2810021A19Rik, Cicr, Cir |
Location: |
Chr.2:73283872-73312592 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cir1 | NM_025854.3 | NP_080130.2 | ||
| AK006260.1 | BAB24488.1 | |||
| AK008242.1 | ||||
| AK131674.1 | BAE20758.1 | |||
| BC089602.1 | AAH89602.1 | |||
| BC094283.1 | AAH94283.1 | |||
| CJ050326.1 |
More Information
Set Membership: |
|

Genomic Map