Assay Details
Target Gene Details
Entrez Gene ID: | 69888 |
Gene Name: | cytochrome P450, family 2, subfamily c, polypeptide 66 |
Gene Aliases: |
2010301M18Rik |
Location: |
Chr.19:39113898-39186756 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2c66 | NM_001011707.1 | NP_001011707.1 | ||
| AK008512.2 | ||||
| AY227734.1 | AAP55508.1 | |||
| BC147819.1 | ||||
| BC147824.1 |
More Information
Set Membership: |
|

Genomic Map