Assay Details
Target Gene Details
Entrez Gene ID: | 108857 |
Gene Name: | ankyrin repeat and KH domain containing 1 |
Gene Aliases: |
1110004O12Rik, 4933432B13Rik, 9130019P20Rik, A530027J04Rik, A630021B20Rik, AA571404, Mask, mFLJ00246, mKIAA1085 |
Location: |
Chr.18:36560603-36658908 on Build GRCm38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankhd1 | NM_175375.3 | NP_780584.2 | ||
| AK017018.1 | ||||
| AK029322.1 | BAC26395.1 | |||
| AK042754.1 | BAC31353.1 | |||
| AK131166.1 | BAD21416.1 |
More Information
Set Membership: |
|

Genomic Map