Assay Details
Target Gene Details
Entrez Gene ID: | 22384 |
Gene Name: | eukaryotic translation initiation factor 4H |
Gene Aliases: |
AU018978, D5Ertd355e, E430026L18Rik, Ef4h, Wbscr1, Wscr1, mKIAA0038 |
Location: |
Chr.5:134619876-134639494 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif4h | NM_033561.1 | NP_291039.1 | ||
AK045004.1 | BAC32180.1 | |||
AK088818.1 | BAC40593.1 | |||
AK122194.1 | 4 | 1590 | BAC65476.3 | |
AK132034.1 | BAE20955.1 | |||
AK135390.1 | BAE22517.1 | |||
AK136999.1 | BAE23201.1 | |||
AK149841.1 | BAE29116.1 | |||
AK150088.1 | BAE29297.1 | |||
AK166567.1 | BAE38858.1 | |||
AK168872.1 | BAE40690.1 | |||
AK169312.1 | BAE41067.1 | |||
AK169353.1 | BAE41102.1 | |||
AK169407.1 | BAE41153.1 | |||
AK177188.1 | ||||
AK182256.1 | ||||
AK184202.1 | ||||
AK184510.1 | ||||
AK188319.1 | ||||
AK191530.1 | ||||
AK192457.1 | ||||
AK193051.1 | ||||
AK193532.1 | ||||
AK193739.1 | ||||
AK195570.1 | ||||
AK196646.1 | ||||
AK196800.1 | ||||
AK197761.1 | ||||
AK197946.1 | ||||
AK200353.1 | ||||
AK200533.1 | ||||
AK201461.1 | ||||
AK203547.1 | ||||
AK215628.1 | ||||
BC014796.1 | AAH14796.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
