Assay Details
Target Gene Details
Entrez Gene ID: | 381101 |
Gene Name: | 2'-deoxynucleoside 5'-phosphate N-hydrolase 1 |
Gene Aliases: |
C76683, Rcl |
Location: |
Chr.17:46496789-46499618 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnph1 | NM_207161.3 | NP_997044.2 | ||
| AK146723.1 | BAE27386.1 | |||
| AK151589.1 | BAE30529.1 | |||
| BC048355.1 | AAH48355.1 |
More Information
Set Membership: |
|

Genomic Map