Assay Details
Target Gene Details
| Entrez Gene ID: | 432450 | 
| Gene Name: | Na+/K+ transporting ATPase interacting 2 | 
| Gene Aliases: | 6330571D19Rik, AW455467, Tcba1 | 
| Location: | Chr.10:31689310-32893997 on Build GRCm38 | 
| Assay Gene Location: | Within Intron 4 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Nkain2 | NM_001013411.2 | NP_001013429.2 | ||
| AY940478.1 | AAX33293.1 | |||
| DQ065607.1 | AAY68416.1 | |||
| EF058049.2 | ||||
| EF058050.1 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic