Assay Details
Target Gene Details
Entrez Gene ID: | 66979 |
Gene Name: | polymerase (DNA-directed), epsilon 4 (p12 subunit) |
Gene Aliases: |
2400007P05Rik, 5830430F06Rik |
Location: |
Chr.6:82618925-82652936 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pole4 | NM_025882.3 | NP_080158.1 | ||
| AK010135.1 | BAB26722.1 | |||
| AK010300.1 | BAB26833.1 | |||
| AK011595.1 | BAB27723.1 | |||
| AK017956.1 | ||||
| AK145365.1 | ||||
| AK160582.1 | BAE35889.1 | |||
| AK167631.1 | BAE39682.1 | |||
| BC023189.1 | AAH23189.1 |
More Information
Set Membership: |
|

Genomic Map