Assay Details
Target Gene Details
Entrez Gene ID: | 218699 |
Gene Name: | PX domain containing serine/threonine kinase |
Gene Aliases: |
C230080L11Rik, D14Ertd813e, MONaKA |
Location: |
Chr.14:8098191-8165117 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pxk | NM_145458.3 | NP_663433.2 | ||
NM_178279.2 | NP_840063.1 | |||
AK028501.1 | ||||
AK048910.1 | BAC33489.1 | |||
AK147037.1 | ||||
AK150337.1 | BAE29479.1 | |||
AK150413.1 | BAE29537.1 | |||
AK150660.1 | ||||
AK151098.1 | ||||
AK152058.1 | BAE30914.1 | |||
AK152789.1 | BAE31498.1 | |||
AK153067.1 | BAE31693.1 | |||
AK156146.1 | BAE33603.1 | |||
AK157364.1 | BAE34065.1 | |||
AK166036.1 | ||||
AK169112.1 | ||||
AK169565.1 | BAE41231.1 | |||
AK170361.1 | BAE41744.1 | |||
AK172347.1 | BAE42959.1 | |||
AK172463.1 | ||||
BC016131.1 | AAH16131.1 | |||
DQ095196.1 | AAZ04664.1 | |||
DQ095197.1 | AAZ04665.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
