Assay Details
Target Gene Details
Entrez Gene ID: | 219022 |
Gene Name: | tetratricopeptide repeat domain 5 |
Gene Aliases: |
- |
Location: |
Chr.14:50765409-50785520 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc5 | NM_001080949.2 | NP_001074418.1 | ||
| NM_177625.4 | NP_808293.1 | |||
| AK044781.1 | BAC32087.1 | |||
| AK077874.1 | ||||
| AK081214.1 | BAC38166.1 | |||
| AK133953.1 | ||||
| AK157777.1 | BAE34192.1 | |||
| AK161279.1 | BAE36290.1 | |||
| BC003272.1 | AAH03272.1 | |||
| BC025610.1 | AAH25610.1 | |||
| BC092074.1 | AAH92074.1 | |||
| BY278375.1 |
More Information
Set Membership: |
|

Genomic Map