Assay Details
Target Gene Details
Entrez Gene ID: | 70408 |
Gene Name: | polymerase (RNA) III (DNA directed) polypeptide F |
Gene Aliases: |
2810411G20Rik, 3010019O03Rik, 3110032A07Rik, RPC39, RPC6 |
Location: |
Chr.2:144527745-144541779 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr3f | NM_029763.3 | NP_084039.2 | ||
| AK013084.1 | BAB28638.1 | |||
| AK013897.1 | ||||
| AK014110.1 | ||||
| AK029286.1 | BAC26372.1 | |||
| AK031473.1 | 1 | 508 | ||
| AK036166.1 | BAC29327.1 | |||
| AK076538.1 | BAC36385.1 | |||
| AK144679.1 | BAE26008.1 | |||
| AK171146.1 | 1 | 510 | ||
| BC009159.1 | AAH09159.1 |
More Information
Set Membership: |
|

Genomic Map