Assay Details
Target Gene Details
Entrez Gene ID: | 80886 |
Gene Name: | SUMO/sentrin specific peptidase 3 |
Gene Aliases: |
AA408656, Smt3ip, Smt3ip1 |
Location: |
Chr.11:69673110-69682084 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Senp3 | NM_001163571.1 | NP_001157043.1 | ||
| NM_030702.4 | NP_109627.3 | |||
| AF194031.1 | AAG28418.1 | |||
| AK045071.1 | BAC32209.1 | |||
| AK088608.1 | BAC40451.1 | |||
| AY008764.1 | AAG33253.1 | |||
| BC037014.1 | AAH37014.1 |
More Information
Set Membership: |
|

Genomic Map