Assay Details
Target Gene Details
Entrez Gene ID: | 14489 |
Gene Name: | myotrophin |
Gene Aliases: |
5033418D15Rik, Gcdp, V1 |
Location: |
Chr.6:35508824-35539888 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtpn | NM_008098.4 | NP_032124.1 | ||
| AK002620.1 | BAB22235.1 | |||
| AK028027.1 | BAC25707.1 | |||
| AK049398.1 | BAC33734.1 | |||
| AK083888.1 | BAC39050.1 | |||
| AK132029.1 | BAE20953.1 | |||
| AK145755.1 | BAE26630.1 | |||
| AK146305.1 | BAE27061.1 | |||
| AK146473.1 | BAE27198.1 | |||
| AK150750.1 | BAE29820.1 | |||
| AK151106.1 | BAE30116.1 | |||
| AK151147.1 | BAE30153.1 | |||
| AK151485.1 | BAE30438.1 | |||
| AK151564.1 | BAE30506.1 | |||
| AK151640.1 | BAE30570.1 | |||
| AK151687.1 | BAE30611.1 | |||
| AK152272.1 | BAE31088.1 | |||
| AK152317.1 | ||||
| BC043084.1 | AAH43084.1 | |||
| BC054811.1 | AAH54811.1 | |||
| CA559569.1 | ||||
| D78188.1 | ||||
| U20290.1 | AAA86719.1 |
More Information
Set Membership: |
|

Genomic Map