Assay Details
Target Gene Details
Entrez Gene ID: | 272009 |
Gene Name: | serine/arginine-rich splicing factor 12 |
Gene Aliases: |
B830039L16, Sfrs13b, Srrp, Srsf13b |
Location: |
Chr.4:33208987-33233344 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srsf12 | NM_177774.4 | NP_808442.2 | ||
| AK046869.1 | BAC32901.1 |
More Information
Set Membership: |
|

Genomic Map