Assay Details
Target Gene Details
Entrez Gene ID: | 76485 |
Gene Name: | glycosyltransferase 8 domain containing 1 |
Gene Aliases: |
2410004H05Rik, 5430414N14Rik, AI450005 |
Location: |
Chr.14:31001347-31012441 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glt8d1 | NM_001165930.1 | NP_001159402.1 | ||
| NM_029626.2 | NP_083902.2 | |||
| AK010388.1 | BAB26903.1 | |||
| AK077388.1 | ||||
| AK160461.1 | BAE35802.1 | |||
| BC055709.1 | AAH55709.1 | |||
| BC058731.1 | AAH58731.1 | |||
| BC069873.1 | AAH69873.1 | |||
| CX209391.1 |
More Information
Set Membership: |
|

Genomic Map