Assay Details
Target Gene Details
Entrez Gene ID: | 14645 |
Gene Name: | glutamate-ammonia ligase (glutamine synthetase) |
Gene Aliases: |
GS, Glns |
Location: |
Chr.1:153899929-153909723 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glul | NM_008131.4 | NP_032157.2 | ||
| AK137969.1 | ||||
| AK159106.1 | BAE34822.1 | |||
| AK160670.1 | BAE35950.1 | |||
| AK162685.1 | BAE37022.1 | |||
| AK168493.1 | BAE40380.1 | |||
| AY044241.1 | AAK95328.1 | |||
| BC015086.1 | AAH15086.1 | |||
| U09114.1 | AAA17989.1 | |||
| X16314.1 | CAA34381.1 |
More Information
Set Membership: |
|

Genomic Map