Assay Details
Target Gene Details
Entrez Gene ID: | 104318 |
Gene Name: | casein kinase 1, delta |
Gene Aliases: |
1200006A05Rik, AA409348, D930010H05Rik |
Location: |
Chr.11:120958010-120991333 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csnk1d | NM_027874.2 | NP_082150.1 | ||
| NM_139059.2 | NP_620690.1 | |||
| AK004606.1 | BAB23405.1 | |||
| AK051052.1 | ||||
| AK088642.1 | BAC40472.1 | |||
| AK140317.1 | BAE24329.1 | |||
| AK143534.1 | ||||
| AK146600.1 | BAE27294.1 | |||
| AK150387.1 | ||||
| AK150815.1 | BAE29878.1 | |||
| AK152721.1 | BAE31444.1 | |||
| AK157812.1 | BAE34206.1 | |||
| AK182511.1 | ||||
| AK210923.1 | ||||
| BC004604.1 | AAH04604.1 |
More Information
Set Membership: |
|

Genomic Map