Assay Details
Target Gene Details
Entrez Gene ID: | 66136 |
Gene Name: | zinc ribbon domain containing, 1 |
Gene Aliases: |
1110014N07Rik, Rpa12 |
Location: |
Chr.17:36954349-36958520 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Znrd1 | NM_023162.4 | NP_075651.1 | ||
| AF230339.1 | AAG50161.1 | |||
| AK003714.1 | BAB22954.1 | |||
| AK005976.1 | BAB24350.1 | |||
| AK019152.1 | ||||
| BC043016.1 | AAH43016.1 |
More Information
Set Membership: |
|

Genomic Map