Assay Details
Target Gene Details
Entrez Gene ID: | 78294 |
Gene Name: | ribosomal protein S27A |
Gene Aliases: |
0610006J14Rik, Uba52, Ubb, Ubc |
Location: |
Chr.11:29545842-29548040 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps27a | NM_001033865.1 | NP_001029037.1 | ||
| NM_024277.2 | NP_077239.1 | |||
| AK018706.1 | BAB31357.1 | |||
| AK159742.1 | BAE35335.1 | |||
| BC002108.1 | AAH02108.1 | |||
| BC081446.1 | AAH81446.1 | |||
| BC096392.1 | AAH96392.1 |
More Information
Set Membership: |
|

Genomic Map