Assay Details
Target Gene Details
Entrez Gene ID: | 19982 |
Gene Name: | ribosomal protein L36A |
Gene Aliases: |
L44L, Rpl44 |
Location: |
Chr.X:134585654-134588062 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl36a | NM_019865.5 | NP_063918.1 | ||
| AK002540.1 | BAB22175.1 | |||
| AK012502.1 | BAB28285.1 | |||
| AK164483.1 | 1 | 133 | ||
| AK168535.1 | BAE40413.1 | |||
| AK170127.1 | BAE41582.1 | |||
| AV453514.1 | ||||
| BC027515.1 | AAH27515.1 | |||
| BC086777.1 | AAH86777.1 | |||
| BU529569.1 |
More Information
Set Membership: |
|

Genomic Map