Assay Details
Target Gene Details
Entrez Gene ID: | 72303 |
Gene Name: | cytochrome P450, family 2, subfamily c, polypeptide 65 |
Gene Aliases: |
2210009K14Rik |
Location: |
Chr.19:39061006-39093948 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2c65 | NM_028191.2 | NP_082467.2 | ||
| AK008688.1 | BAB25834.1 | |||
| AY227733.1 | AAP55507.1 | |||
| BC117531.1 | ||||
| BC118514.1 |
More Information
Set Membership: |
|

Genomic Map