Assay Details
Target Gene Details
Entrez Gene ID: | 241118 |
Gene Name: | acid-sensing (proton-gated) ion channel family member 4 |
Gene Aliases: |
Accn4, BNAC4 |
Location: |
Chr.1:75450510-75474340 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asic4 | NM_183022.3 | NP_898843.1 | ||
| BC046481.1 | AAH46481.1 | |||
| BC055772.1 | AAH55772.1 |
More Information
Set Membership: |
|

Genomic Map