Assay Details
Target Gene Details
Entrez Gene ID: | 404195 |
Gene Name: | cytochrome P450, family 2, subfamily c, polypeptide 54 |
Gene Aliases: |
EG404195 |
Location: |
Chr.19:40037940-40073845 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2c54 | NM_206537.2 | NP_996260.1 | ||
| AI181879.1 | ||||
| BF659738.1 |
More Information
Set Membership: |
|

Genomic Map