Assay Details
Target Gene Details
Entrez Gene ID: | 19039 |
Gene Name: | lectin, galactoside-binding, soluble, 3 binding protein |
Gene Aliases: |
90K, CyCAP, MAC-2BP, Ppicap, Tango10b |
Location: |
Chr.11:118392752-118401931 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lgals3bp | NM_011150.2 | NP_035280.1 | ||
| AK079915.1 | BAC37782.1 | |||
| AK137836.1 | BAE23505.1 | |||
| AK150021.1 | BAE29246.1 | |||
| AK152182.1 | BAE31013.1 | |||
| AK153313.1 | BAE31894.1 | |||
| AK153396.1 | BAE31958.1 | |||
| AK155031.1 | BAE33003.1 | |||
| BC090658.1 | ||||
| L16894.1 | AAA37499.1 | |||
| X67809.1 | CAA48010.1 |
More Information
Set Membership: |
|

Genomic Map