Assay Details
Target Gene Details
Entrez Gene ID: | 16069 |
Gene Name: | immunoglobulin joining chain |
Gene Aliases: |
9530090F24Rik, AI323815, Igj, Jch |
Location: |
Chr.5:88519809-88527897 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jchain | NM_152839.3 | NP_690052.2 | ||
| AB644392.1 | ||||
| AK078865.1 | BAC37428.1 | |||
| AK160344.1 | BAE35749.1 | |||
| BC006026.1 | AAH06026.1 | |||
| J00544.1 | AAA38673.1 | |||
| M90766.1 | AAA37918.1 |
More Information
Set Membership: |
|

Genomic Map