Assay Details
Target Gene Details
Entrez Gene ID: | 74577 |
Gene Name: | galactosidase, beta 1-like |
Gene Aliases: |
4833408P15Rik |
Location: |
Chr.1:75198235-75210816 on Build GRCm38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glb1l | NM_029010.1 | NP_083286.1 | ||
| AK014667.1 | BAB29494.1 | |||
| AK029925.1 | BAC26681.1 | |||
| AK170689.1 | 1 | 389 | ||
| BC021773.1 | AAH21773.1 |
More Information
Set Membership: |
|

Genomic Map