Assay Details
Target Gene Details
Entrez Gene ID: | 218639 |
Gene Name: | ADP-ribosylation factor-like 15 |
Gene Aliases: |
A430036I03, Arfrp2, C230032K13Rik |
Location: |
Chr.13:113794506-114157462 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arl15 | NM_172595.4 | NP_766183.1 | ||
| AK039965.1 | BAC30485.1 | |||
| AK048736.1 | BAC33440.1 | |||
| AK050979.1 | BAE20688.1 | |||
| AK052164.1 | BAC34864.1 | |||
| AK053953.1 | ||||
| BC120664.1 | ||||
| BC120666.1 | ||||
| BC144993.1 | ||||
| CJ063085.1 | ||||
| CJ066332.1 |
More Information
Set Membership: |
|

Genomic Map